Paracetamol (Acetaminophen) 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-837
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:103-90-2
- Description:Paracetamol (Acetaminophen) 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₈H₉NO₂
- Molecular Weight:151.16 g/mol
- Cat. No.:77696-837