Heroin-D9 (Diacetylmorphine-D9) 1.0 mg/ml in Acetonitrile, LoGiCal, LGC Standards
Catalog # 77696-836
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1338713-49-7
- Description:Heroin-D9 (Diacetylmorphine-D9) 1.0 mg/ml in Acetonitrile
- Size:1 ml
- Formula:C₂₁H₁₄D₉NO₅
- Cat. No.:77696-836