Morphine 0.1 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-834
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:57-27-2
- Description:Morphine 0.1 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₇H₁₉NO₃
- Molecular Weight:285.34 g/mol
- Cat. No.:77696-834