Ibuprofen 1.0 mg/ml in Methanol, LoGiCal, LGC Standards
Catalog # 77696-818
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:15687-27-1
- Description:Ibuprofen 1.0 mg/ml in Methanol
- Size:1 ml
- Formula:C₁₃H₁₈O₂
- Molecular Weight:206.3 g/mol
- Cat. No.:77696-818