N-Nitroso-di-n-butylamine 1000 µg/mL in Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-539
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:924-16-3
- Description:N-Nitroso-di-n-butylamine 1000 µg/mL in Methanol
- Size:1 ml
- Formula:C₈H₁₈N₂O
- Molecular Weight:158.25 g/mol
- Cat. No.:77696-539