Permethrin 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-510
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52645-53-1
- Description:Permethrin 100 µg/mL in Cyclohexane
- Size:1 ml
- Formula:C₂₁H₂₀Cl₂O₃
- Molecular Weight:391.3 g/mol
- Cat. No.:77696-510