Paclobutrazol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-507
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:76738-62-0
- Description:Paclobutrazol 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₅H₂₀ClN₃O
- Molecular Weight:293.795 g/mol
- Cat. No.:77696-507