Malathion 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-493
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:121-75-5
- Description:Malathion 100 µg/mL in Cyclohexane
- Size:1 ml
- Formula:C₁₀H₁₉O₆PS₂
- Cat. No.:77696-493