n-Heptyltin trichloride 100 µg/mL in Methyl-tert-butyl ether, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-482
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:59344-47-7
- Description:n-Heptyltin trichloride 100 µg/mL in Methyl-tert-butyl ether
- Size:1 ml
- Formula:C₇H₁₅Cl₃Sn
- Cat. No.:77696-482