Dinoseb 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-458
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:88-85-7
- Description:Dinoseb 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₀H₁₂N₂O₅
- Molecular Weight:240.22 g/mol
- Cat. No.:77696-458