$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Simazine 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
Simazine 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
77696-411
:
LGC Standards
New Product
:
Simazine 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
77696-411
:
LGC Standards
:
DRE-GA09011128AC
New Product
:
CAS No.:
122-34-9
Description:
Simazine 100 µg/mL in Acetone
Size:
1 ml
Formula:
C₇H₁₂ClN₅
Molecular Weight:
201.66 g/mol
Cat. No.:
77696-411