Moxidectin, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-364
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:113507-06-5
- Description:Moxidectin
- Size:25 mg
- Formula:C₃₇H₅₃NO₈
- Molecular Weight:639.83 g/mol
- Cat. No.:77696-364