n-Octyltin-trichloride, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-327
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:3091-25-6
- Description:n-Octyltin-trichloride
- Size:100 mg
- Formula:C₈H₁₇Cl₃Sn
- Cat. No.:77696-327