Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Puerto Rico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
2-Ethyl-1,3-hexandiol, Dr. Ehrenstorfer, LGC Standards
2-Ethyl-1,3-hexandiol, Dr. Ehrenstorfer, LGC Standards
77678-005
:
LGC Standards
New Product
:
2-Ethyl-1,3-hexandiol, Dr. Ehrenstorfer, LGC Standards
77678-005
:
LGC Standards
:
DRE-C13340000
New Product
:
CAS No.:
94-96-2
Description:
2-Ethyl-1,3-hexandiol
Size:
250 mg
Formula:
C₈H₁₈O₂
Molecular Weight:
146.23 g/mol
Cat. No.:
77678-005