$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Ethanol, Dr. Ehrenstorfer, LGC Standards
Ethanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-321
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
Ethanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-321
Supplier:
LGC Standards
Supplier Number:
DRE-C13223000-10ML
New Product
CAS Number:
Specifications
CAS No.:
64-17-5
Description:
Ethanol
Size:
10 ml
Formula:
C₂H₅OH
Molecular Weight:
46.07 g/mol
Cat. No.:
77696-321
Specifications
About this item
Product Details
Product Details