Pyribencarb 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-285
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:799247-52-2
- Description:Pyribencarb 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₈H₂₀ClN₃O₃
- Molecular Weight:361.82 g/mol
- Cat. No.:77696-285