Propionic acid 1000 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-282
New Product
:
- CAS No.:79-09-4
- Description:Propionic acid 1000 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₃H₆O₂
- Molecular Weight:74.08 g/mol
- Cat. No.:77696-282