PBDE 209 50 µg/mL in Nonane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-249
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:1163-19-5
- Description:PBDE 209 50 µg/mL in Nonane
- Size:1 ml
- Formula:C₁₂Br₁₀O
- Cat. No.:77696-249