Patent Blue V 100 µg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer, LGC Standards
77696-240
New Product
:
- CAS No.:20262-76-4
- Description:Patent Blue V 100 µg/mL in Acetonitrile:Methanol
- Size:1 ml
- Formula:C₂₇H₃₁N₂NaO₇S₂
- Molecular Weight:582.67 g/mol
- Cat. No.:77696-240