Ochratoxin A 10 µg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer, LGC Standards
77696-228
New Product
:
- CAS No.:303-47-9
- Description:Ochratoxin A 10 µg/mL in Acetonitrile:Methanol
- Size:1 ml
- Formula:C₂₀H₁₈ClNO₆
- Molecular Weight:403.8 g/mol
- Cat. No.:77696-228