Norethisterone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-225
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:68-22-4
- Description:Norethisterone 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₀H₂₆O₂
- Molecular Weight:298.426 g/mol
- Cat. No.:77696-225