Nimodipine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-213
New Product
:
- CAS No.:66085-59-4
- Description:Nimodipine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₁H₂₆N₂O₇
- Molecular Weight:418.45 g/mol
- Cat. No.:77696-213