Nicotinic acid 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-211
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:59-67-6
- Description:Nicotinic acid 10 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₆H₅NO₂
- Molecular Weight:123.11 g/mol
- Cat. No.:77696-211