$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Puerto Rico
ContactUsLinkComponent
Standards
Organic Standards
Naproxen 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Naproxen 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-203
:
LGC Standards
New Product
:
Naproxen 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-203
:
LGC Standards
:
DRE-A15483500AL-100
New Product
:
CAS No.:
22204-53-1
Description:
Naproxen 100 µg/mL in Acetonitrile
Size:
1 ml
Formula:
C₁₄H₁₄O₃
Molecular Weight:
230.26 g/mol
Cat. No.:
77696-203