Moxidectin 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-200
New Product
:
- CAS No.:113507-06-5
- Description:Moxidectin 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₃₇H₅₃NO₈
- Molecular Weight:639.83 g/mol
- Cat. No.:77696-200