N-Methylperfluorooctanesulfonamide 100 µg/mL in Methanol/Water, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-191
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:31506-32-8
- Description:N-Methylperfluorooctanesulfonamide 100 µg/mL in Methanol/Water
- Size:1 ml
- Formula:C₉H₄F₁₇NO₂S
- Molecular Weight:513.18 g/mol
- Cat. No.:77696-191