2-Methyl-2-butanol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-651
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:75-85-4
- Description:2-Methyl-2-butanol 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₅H₁₂O
- Molecular Weight:88.15 g/mol
- Cat. No.:77691-651