Metacycline hydrochloride 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-186
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:3963-95-9
- Description:Metacycline hydrochloride 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₂H₂₃ClN₂O₈
- Molecular Weight:478.88 g/mol
- Cat. No.:77696-186