Megestrol-17-acetate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-173
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:595-33-5
- Description:Megestrol-17-acetate 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₄H₃₂O₄
- Molecular Weight:384.52 g/mol
- Cat. No.:77696-173