Mefentrifluconazole 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-172
New Product
:
- CAS No.:1417782-03-6
- Description:Mefentrifluconazole 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₈H₁₅ClF₃N₃O₂
- Molecular Weight:397.78 g/mol
- Cat. No.:77696-172