Medroxyprogesterone-17-acetate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-171
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:71-58-9
- Description:Medroxyprogesterone-17-acetate 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₄H₃₄O₄
- Molecular Weight:386.53 g/mol
- Cat. No.:77696-171