Levonorgestrel 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-161
New Product
:
- CAS No.:797-63-7
- Description:Levonorgestrel 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₁H₂₈O₂
- Molecular Weight:312.45 g/mol
- Cat. No.:77696-161