Lamotrigine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-158
New Product
:
- CAS No.:84057-84-1
- Description:Lamotrigine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₉H₇Cl₂N₅
- Molecular Weight:256.09 g/mol
- Cat. No.:77696-158