Ketoprofen 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-155
New Product
:
- CAS No.:22071-15-4
- Description:Ketoprofen 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₆H₁₄O₃
- Molecular Weight:254.29 g/mol
- Cat. No.:77696-155