Iopromid 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-152
New Product
:
- CAS No.:73334-07-3
- Description:Iopromid 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₈H₂₄I₃N₃O₈
- Molecular Weight:791.11 g/mol
- Cat. No.:77696-152