Fluticasone propionate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-119
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:80474-14-2
- Description:Fluticasone propionate 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₅H₃₁F₃O₅S
- Molecular Weight:500.57 g/mol
- Cat. No.:77696-119