Fluroxypyr-2-hydroxy 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-116
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:94133-62-7
- Description:Fluroxypyr-2-hydroxy 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₅H₃Cl₂FN₂O
- Molecular Weight:196.99 g/mol
- Cat. No.:77696-116