Fluoxetine hydrochloride 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-112
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:56296-78-7
- Description:Fluoxetine hydrochloride 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₇H₁₈F₃NO·HCl
- Molecular Weight:345.79 g/mol
- Cat. No.:77696-112