Flonicamid (free acid) 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-097
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:158063-66-2
- Description:Flonicamid (free acid) 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₇H₄F₃NO₂
- Molecular Weight:191.11 g/mol
- Cat. No.:77696-097