Felodipine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-088
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:72509-76-3
- Description:Felodipine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₈H₁₉Cl₂NO₄
- Molecular Weight:384.25 g/mol
- Cat. No.:77696-088