Fast Green FCF 100 µg/mL in Acetonitrile/Water, Dr. Ehrenstorfer, LGC Standards
77696-087
New Product
:
- CAS No.:2353-45-9
- Description:Fast Green FCF 100 µg/mL in Acetonitrile/Water
- Size:1 ml
- Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
- Molecular Weight:808.86 g/mol
- Cat. No.:77696-087