Erythrosin B disodium 100 µg/mL in Acetonitrile/Water, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-081
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:16423-68-0
- Description:Erythrosin B disodium 100 µg/mL in Acetonitrile/Water
- Size:1 ml
- Formula:C₂₀H₆I₄Na₂O₅
- Molecular Weight:879.86 g/mol
- Cat. No.:77696-081