Doxycycline hyclate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-076
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:24390-14-5
- Description:Doxycycline hyclate 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₂₂H₂₄N₂O₈·HCl·0.5H₂O·0.5C₂H₆O
- Molecular Weight:512.94 g/mol (503.94 anhy)
- Cat. No.:77696-076