Disperse Red 11 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77696-066
New Product
:
- CAS No.:2872-48-2
- Description:Disperse Red 11 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₅H₁₂N₂O₃
- Molecular Weight:268.3 g/mol
- Cat. No.:77696-066