Disperse Blue 3 100 µg/mL in Acetonitrile:Methanol, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-058
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:2475-46-9
- Description:Disperse Blue 3 100 µg/mL in Acetonitrile:Methanol
- Size:1 ml
- Formula:C₁₇H₁₆N₂O₃
- Molecular Weight:296.32 g/mol
- Cat. No.:77696-058