1,4-Dioxane D8 1000 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-622
New Product
:
- CAS No.:17647-74-4
- Description:1,4-Dioxane D8 1000 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₄D₈O₂
- Molecular Weight:96.04 g/mol
- Cat. No.:77691-622