Lambda-cyhalothrin 1000 µg/mL in Toluene, Dr. Ehrenstorfer, LGC Standards
Catalog # 77696-051
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:91465-08-6
- Description:Lambda-cyhalothrin 1000 µg/mL in Toluene
- Size:1 ml
- Formula:C₂₃H₁₉ClF₃NO₃
- Cat. No.:77696-051