Crotonaldehyde-2,4-dinitrophenylhydrazone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77691-583
New Product
:
- CAS No.:1527-96-4
- Description:Crotonaldehyde-2,4-dinitrophenylhydrazone 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₀H₁₀N₄O₄
- Molecular Weight:250.21 g/mol
- Cat. No.:77691-583