2-Butanol 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-505
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:78-92-2
- Description:2-Butanol 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:H₃CCH₂CH(OH)CH₃
- Molecular Weight:74.12 g/mol
- Cat. No.:77691-505