Benzene 30 µg/mL in N,N-Dimethylacetamide, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-481
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:71-43-2
- Description:Benzene 30 µg/mL in N,N-Dimethylacetamide
- Size:1 ml
- Formula:C₆H₆
- Molecular Weight:78.11 g/mol
- Cat. No.:77691-481